Skip to Content
Merck
All Photos(1)

Key Documents

S-061

Supelco

XLR-11 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28FNO
CAS Number:
Molecular Weight:
329.45
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CN(CCCCCF)C2=C1C=CC=C2)C3C(C)(C)C3(C)C

InChI

1S/C21H28FNO/c1-20(2)19(21(20,3)4)18(24)16-14-23(13-9-5-8-12-22)17-11-7-6-10-15(16)17/h6-7,10-11,14,19H,5,8-9,12-13H2,1-4H3

InChI key

PXLDPUUMIHVLEC-UHFFFAOYSA-N

General description

XLR-11 is a synthetic cannabinoid of the tetramethylcyclopropyl series found in newer Spice blends. Along with UR-144, cyclopropyl Spice analogs have seen wide popularity in the illicit markets since late 2012. XLR-11 is similar in structure to UR-144 and A-796,260, two compounds originally developed as pharmaceutical drug candidates due to their high affinity for the cannabinoid receptor CB2.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service