Skip to Content
Merck
All Photos(1)

Key Documents

900200O

Avanti

Dolichol (13~21)

Avanti Research - A Croda Brand 900200O

Synonym(s):

Dolichol Mixture (13~21)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mg (900200O-1mg)
pkg of 1 × 2 mg (900200O-2mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900200O

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)=CCC/C(C)=C/CC/C(C)=C/CC/C(C)=C\CC[C@@H](CCO)C

General description

Dolichol is composed of repeated units of isoprenoids and forms a long chain unsaturated molecule. It is known to be widely present in all eukaryotic cells. Dolichol is predominantly synthesised in endoplasmic reticulum and localized to the phospholipid bilayer.

Biochem/physiol Actions

Dolichol influences the process of protein transfer in the endoplasmic reticulum through protein glycosylation. It acts as a lipid carrier of sugars.

Packaging

5 mL Clear Glass Sealed Ampule (900200O-1mg)
5 mL Clear Glass Sealed Ampule (900200O-2mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dolichol biosynthesis and its effects on the unfolded protein response and abiotic stress resistance in Arabidopsis
Zhang H, et al.
Plant Cell, 20(7), 1879-1898 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service