GF55219484
Tungsten-copper alloy, W60Cu40
175mm tube, 3.15mm outside diameter, 1.15mm wall thickness, 0.85mm inside diameter
Synonym(s):
W 187300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 552-194-84
L × wall thickness
175 mm × 1.15 mm
O.D. × I.D.
3.15 mm × 0.85 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service