Skip to Content
Merck
All Photos(1)

Key Documents

907820

Sigma-Aldrich

Platinum on carbon

Evonik Noblyst® P8075 1% Pt

Synonym(s):

Pt/C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600

form

powder

contains

~50% water

extent of labeling

1 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

activated carbon support

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Evonik Noblyst® P8075 1% Pt is a platinum on carbon catalyst. It has been shown to be effective in the following reactions:
  • hydrogenation of aromatic nitro groups in halonitroaromatics
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Other Notes

Unit weight is of net dry catalyst.

Legal Information

Manufactured by Evonik Degussa GmbH.
Noblyst is a registered trademark of Evonik Degussa GmbH

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

An application guide to help determine which heterogeneous catalysts are best for different synthetic applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service