Skip to Content
Merck
All Photos(2)

Key Documents

768979

Sigma-Aldrich

Ethane

Messer® CANGas, 99.95%

Synonym(s):

Ethane gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH3
CAS Number:
Molecular Weight:
30.07
Beilstein:
1730716
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.05 (vs air)

Quality Level

vapor pressure

37.95 atm ( 21.1 °C)

description

Filling mass: 17g
Filling volume: 12L

Assay

99.95%

form

gas

autoignition temp.

881 °F

expl. lim.

13 %

bp

−88 °C (lit.)

mp

−172 °C (lit.)

density

0.362 g/mL at 20 °C (lit.)

SMILES string

CC

InChI

1S/C2H6/c1-2/h1-2H3

InChI key

OTMSDBZUPAUEDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethane gas is colourless, odourless, and narcotic. It is a liquefied gas that is lighter than air. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-211.0 °F - closed cup

Flash Point(C)

-135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service