Skip to Content
Merck
All Photos(1)

Key Documents

588040

Sigma-Aldrich

Chloroethane-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CH2Cl
CAS Number:
Molecular Weight:
65.51
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

bp

12.3 °C (lit.)

mass shift

M+1

SMILES string

C[13CH2]Cl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3/i2+1

InChI key

HRYZWHHZPQKTII-VQEHIDDOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-58.0 °F - closed cup

Flash Point(C)

-50 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service