HPLC Analysis of pBr322 DNA-Hae III Digest on TSKgel® DEAE-NPR
Materials
analytical column
Product No.
Description
Pricing
TSKgel® DEAE-NPR HPLC Column
phase DEAE (diethylaminoethyl), L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle sizeCONDITIONS
column
TSKgel DEAE-NPR, 3.5 cm × 4.6 mm I.D., 2.5 μm particles (with 5 mm DEAE-NPR guard column) (813075)
mobile phase
[A] 1 M sodium chloride/25 mm Tris-HCl, pH 9.0; [B] 25 mM Tris-HCl, pH 9.0
flow rate
1 mL/min
column temp.
ambient
detector
UV, 260 nm
injection
20 μL of 50 μg/mL Hae III digest of pBR322 DNA
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation