GC Analysis of Nitrosamines in Water on PTA-5 after SPME using 65 μm PDMS/DVB Fiber
![GC Analysis of Nitrosamines in Water on PTA-5 after SPME using 65 μm PDMS/DVB Fiber suitable for GC, application for SPME](/deepweb/assets/sigmaaldrich/product/images/238/216/1d2cbfae-d8a8-461c-8ca0-5176575cb608/800/1d2cbfae-d8a8-461c-8ca0-5176575cb608.jpg)
Materials
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holderCONDITIONS
sample/matrix
analytes in water +25% KCl, pH 10.0
SPME fiber
65 μm polydimethylsiloxane/divinylbenzene (57310-U)
extraction
immersion, 15 min (rapid stirring)
desorption process
270 °C for 1 min
column
PTA-5, 30 m × 0.32 mm I.D., 0.5 μm (24331)
oven
50 °C (1 min), 10 °C/min to 250 °C (2 min)
inj. temp.
250 °C
detector
MSD, SIM
carrier gas
helium, 30 cm/sec
liner
0.75 mm I.D., SPME