GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 85 μm Polyacrylate Fiber, Fast GC Analysis
![GC Analysis of Basic Drugs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 85 μm Polyacrylate Fiber, Fast GC Analysis suitable for GC, application for SPME](/deepweb/assets/sigmaaldrich/product/images/374/297/c2d9b93a-75f3-4c36-b87b-126cebd0843f/800/c2d9b93a-75f3-4c36-b87b-126cebd0843f.jpg)
Materials
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly polyacrylate (PA)
df 85 μm(PA, for use with manual holder, needle size 24 gaCONDITIONS
sample/matrix
500 ug/L of each compound in 2 mL 0.05M phosphate buffer, pH=11, plus sodium chloride (NaCl) to 25% (w/v)
SPME fiber
85 μm Polyacrylate (57304)
extraction
immersion, 450 rpm stirring at room temp. for 15 min
desorption process
250 °C for 3 min
column
SLB-5ms, 20 m x 0.18 mm I.D. x 0.36 μm (28576-U)
oven
100 °C (1 min), 20 °C/min to 325 °C (5 min)
MSD interface
325 °C
carrier gas
helium, 1.5 mL/min. constant
liner
0.75 mm I.D. SPME
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany