Merck
  • IL10 Release upon PD-1 Blockade Sustains Immunosuppression in Ovarian Cancer.
IL10 Release upon PD-1 Blockade Sustains Immunosuppression in Ovarian Cancer.
Cancer research (2017-10-11)
Purushottam Lamichhane, Lavakumar Karyampudi, Barath Shreeder, James Krempski, Deborah Bahr, Joshua Daum, Kimberly R Kalli, Ellen L Goode, Matthew S Block, Martin J Cannon, Keith L Knutson
ABSTRACT
Ligation of programmed cell death-1 (PD-1) in the tumor microenvironment is known to inhibit effective adaptive antitumor immunity. Blockade of PD-1 in humans has resulted in impressive, durable regression responses in select tumor types. However, durable responses have been elusive in ovarian cancer patients. PD-1 was recently shown to be expressed on and thereby impair the functions of tumor-infiltrating murine and human myeloid dendritic cells (TIDC) in ovarian cancer. In the present work, we characterize the regulation of PD-1 expression and the effects of PD-1 blockade on TIDC. Treatment of TIDC and bone marrow-derived dendritic cells (DC) with IL10 led to increased PD-1 expression. Both groups of DCs also responded to PD-1 blockade by increasing production of IL10. Similarly, treatment of ovarian tumor-bearing mice with PD-1 blocking antibody resulted in an increase in IL10 levels in both serum and ascites. While PD-1 blockade or IL10 neutralization as monotherapies were inefficient, combination of these two led to improved survival and delayed tumor growth; this was accompanied by augmented antitumor T- and B-cell responses and decreased infiltration of immunosuppressive MDSC. Taken together, our findings implicate compensatory release of IL10 as one of the adaptive resistance mechanisms that undermine the efficacy of anti-PD-1 (or anti-PD-L1) monotherapies and prompt further studies aimed at identifying such resistance mechanisms.
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3