Merck
  • Human mesenchymal stromal cells transplanted into mice stimulate renal tubular cells and enhance mitochondrial function.
Human mesenchymal stromal cells transplanted into mice stimulate renal tubular cells and enhance mitochondrial function.
Nature communications (2017-10-19)
Luca Perico, Marina Morigi, Cinzia Rota, Matteo Breno, Caterina Mele, Marina Noris, Martino Introna, Chiara Capelli, Lorena Longaretti, Daniela Rottoli, Sara Conti, Daniela Corna, Giuseppe Remuzzi, Ariela Benigni
ABSTRACT
Mesenchymal stromal cells (MSCs) are renoprotective and drive regeneration following injury, although cellular targets of such an effect are still ill-defined. Here, we show that human umbilical cord (UC)-MSCs transplanted into mice stimulate tubular cells to regain mitochondrial mass and function, associated with enhanced microtubule-rich projections that appear to mediate mitochondrial trafficking to create a reparative dialogue among adjacent tubular cells. Treatment with UC-MSCs in mice with cisplatin-induced acute kidney injury (AKI) regulates mitochondrial biogenesis in proximal tubuli by enhancing PGC1α expression, NAD+ biosynthesis and Sirtuin 3 (SIRT3) activity, thus fostering antioxidant defenses and ATP production. The functional role of SIRT3 in tubular recovery is highlighted by data that in SIRT3-deficient mice with AKI, UC-MSC treatment fails to induce renoprotection. These data document a previously unrecognized mechanism through which UC-MSCs facilitate renal repair, so as to induce global metabolic reprogramming of damaged tubular cells to sustain energy supply.Mesenchymal stromal cells drive renal regeneration following injury. Here, the authors show that human mesenchymal stromal cells, when transplanted into mice with acute kidney injury, stimulate renal tubular cell growth and enhance mitochondrial function via SIRT3.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-KIF5C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mitochondria Antibody, clone 113-1, Cy3 Conjugate, clone 113-1, from mouse, CY3 conjugate
Sigma-Aldrich
ANTI-KIF5C-2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, Alexa Fluor 488 conjugate, clone 235-1, from mouse, ALEXA FLUOR 488