Merck
  • Dexamethasone inhibits interleukin-1beta-induced corneal neovascularization: role of nuclear factor-kappaB-activated stromal cells in inflammatory angiogenesis.
Dexamethasone inhibits interleukin-1beta-induced corneal neovascularization: role of nuclear factor-kappaB-activated stromal cells in inflammatory angiogenesis.
The American journal of pathology (2007-08-11)
Shintaro Nakao, Yasuaki Hata, Muneki Miura, Kousuke Noda, Yusuke N Kimura, Shuhei Kawahara, Takeshi Kita, Toshio Hisatomi, Toru Nakazawa, Yiping Jin, M Reza Dana, Michihiko Kuwano, Mayumi Ono, Tatsuro Ishibashi, Ali Hafezi-Moghadam
ABSTRACT
Dexamethasone, a synthetic corticosteroid, is widely used as a potent anti-inflammatory drug in various diseases including corneal angiogenesis. However, dexamethasone's impact on interleukin (IL)-1beta-dependent inflammatory angiogenesis is unknown. Here, we show that dexamethasone inhibits IL-1beta-induced neovascularization and the expression of the angiogenesis-related factors, vascular endothelial growth factor-A, KC, and prostaglandin E(2) in the mouse cornea 2 days after IL-1beta implantation. IL-1beta caused IkappaB-alpha phosphorylation in corneal stromal cells but not in infiltrated CD11b(+) cells 2 days after IL-1beta implantation. In contrast, both cell types were positive for phosphorylated IkappaB-alpha 4 days after IL-1beta implantation. Dexamethasone significantly inhibited IkappaB-alpha phosphorylation 2 and 4 days after IL-1beta implantation. Furthermore, dexamethasone inhibited IL-1beta-induced expression of vascular endothelial growth factor-A, KC, and prostaglandin E(2), and signaling of nuclear factor (NF)-kappaB in corneal fibroblasts in vitro. A selective NF-kappaB inhibitor attenuated IL-1beta-induced corneal angiogenesis. These findings suggest that NF-kappaB activation in the corneal stromal cells is an important early event during IL-1beta-induced corneal angiogenesis and that dexamethasone inhibits IL-1beta-induced angiogenesis partially via blocking NF-kappaB signaling.
Sigma-Aldrich
Mammalian Cell Lysis Kit
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, FITC conjugate, 26 mg/mL, Chemicon®
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture