Merck
  • Evaluation of a novel metal-organic framework as an adsorbent for the extraction of multiclass pesticides from coconut palm (Cocos nucifera L.): An analytical approach using matrix solid-phase dispersion and liquid chromatography.
Evaluation of a novel metal-organic framework as an adsorbent for the extraction of multiclass pesticides from coconut palm (Cocos nucifera L.): An analytical approach using matrix solid-phase dispersion and liquid chromatography.
Journal of separation science (2017-06-24)
Jemmyson Romário de Jesus, Kaline Amaral Wanderley, Severino Alves Júnior, Sandro Navickiene
ABSTRACT
We report the synthesis, characterization, and application of [Zn(1,4-benzenedicarboxylate)(H
Product Description
Supelco
Coumaphos, PESTANAL®, analytical standard
Supelco
Bromopropylate, PESTANAL®, analytical standard
Supelco
Clofentezine, PESTANAL®, analytical standard
Supelco
Teflubenzuron, PESTANAL®, analytical standard
Supelco
Flumetralin, PESTANAL®, analytical standard