Merck
  • New validated high-performance liquid chromatographic method for simultaneous analysis of ten flavonoid aglycones in plant extracts using a C18 fused-core column and acetonitrile-tetrahydrofuran gradient.
New validated high-performance liquid chromatographic method for simultaneous analysis of ten flavonoid aglycones in plant extracts using a C18 fused-core column and acetonitrile-tetrahydrofuran gradient.
Journal of separation science (2012-07-19)
Monika A Olszewska
ABSTRACT
An HPLC method of high resolution has been developed and validated for the simultaneous determination of ten prominent flavonoid aglycones in plant materials using a fused-core C18-silica column (Ascentis® Express, 4.6 mm × 150 mm, 2.7 μm). The separation was accomplished with an acetonitrile-tetrahydrofuran gradient elution at a flow rate of 1 mL/min and temperature of 30°C. UV spectrophotometric detection was employed at 370 nm for flavonols (quercetin [QU], myricetin [MY], isorhamnetin [IS], kaempferol [KA], sexangularetin [SX], and limocitrin [LM]) and 340 nm for flavones (apigenin [AP], acacetin [AC], chrysoeriol [CH], and luteolin [LU]). The high resolution of critical pairs QU/LU (10.50), QU/CH (3.40), AP/CH (2.51), SX/LM (2.30), and IS/KA (2.70) was achieved within 30.3 min. The observed column back pressure was less than 4300 psi, thus acceptable for conventional HPLC equipment. The method was sensitive enough having LODs of 0.115-0.525 ng and good linearity (r > 0.9999) over the test range. The precision values, expressed as RSD values, were <7.5%, and the accuracy was in the range of 95.3-100.2% for all analytes except MY (73.8%). The method was successfully employed for the determination of flavonoids in several medicinal plants, such as Ginkgo biloba, Betula pendula, and a variety of Sorbus species.
Product Description
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 4.6 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® Express C8 Capillary, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® C8 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® C8 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C8 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C8 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Ascentis® Si HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Si HPLC Column, 3 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Si HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Ascentis® Si HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Si HPLC Column, 3 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Si HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Ascentis® Si HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Ascentis® Si HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Si HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 2.1 mm