Merck
  • Second bioluminescence-activating component in the luminous fungus Mycena chlorophos.
Second bioluminescence-activating component in the luminous fungus Mycena chlorophos.
Luminescence : the journal of biological and chemical luminescence (2016-06-09)
Katsunori Teranishi
ABSTRACT
Mycena chlorophos is an oxygen-dependent bioluminescent fungus. The mechanisms underlying its light emission are unknown. A component that increased the bioluminescence intensity of the immature living gills of M. chlorophos was isolated from mature M. chlorophos gills and chemically characterized. The bioluminescence-activating component was found to be trans-3,4-dihydroxycinnamic acid and its bioluminescence activation was highly structure-specific.
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
Hispidin, solid, ≥98% (HPLC)
Sigma-Aldrich
3,5-Dimethoxycinnamic acid, predominantly trans, ≥99%
Supelco
trans-Caffeic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland