Merck
A
Bioconjugate chemistry (2017-06-22)
Renjie Liu, Dharmesh Patel, Hazel R C Screen, C Remzi Becer
ABSTRACT
Electrospun biodegradable membranes have attracted great attention for a range of tissue engineering applications. Among them, poly(ε-caprolactone) (PCL) is one of the most widely used polymers, owing to its well-controlled biocompatibility and biodegradability. However, PCL also has a number of limitations, such as its hydrophobic nature and the lack of functional groups on its side chain, limiting its ability to interact with cells. Herein, we have designed and prepared a series of well-defined A
Sigma-Aldrich
DL-1,2-Isopropylideneglycerol, ≥97.0%
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
2,3,4,5,6-Pentafluorostyrene, 99%, contains 0.1% p-tert-butylcatechol as inhibitor
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%