Merck
  • Tormentic acid inhibits H2O2-induced oxidative stress and inflammation in rat vascular smooth muscle cells via inhibition of the NF-κB signaling pathway.
Tormentic acid inhibits H2O2-induced oxidative stress and inflammation in rat vascular smooth muscle cells via inhibition of the NF-κB signaling pathway.
Molecular medicine reports (2016-08-31)
Yu-Lun Wang, Gen-Yi Sun, Ying Zhang, Jia-Jun He, Shen Zheng, Jing-Na Lin
ABSTRACT
Tormentic acid (TA) is a triterpene isolated from the stem bark of the plant Vochysia divergens and has been reported to exhibit anticancer, anti‑inflammatory and anti‑atherogenic properties. However, the functions of TA in hydrogen peroxide (H2O2)‑induced oxidative stress and inflammation in rat vascular smooth muscle cells (RVSMCs) remain unclear. Therefore, the present study aimed to investigate whether TA suppressed H2O2‑induced oxidative stress and inflammation in RVSMCs, and to determine its molecular mechanisms. The present study demonstrated that TA inhibited reactive oxygen species (ROS) generation, induced H2O2 in RVSMCs, and inhibited H2O2-induced expression of inducible nitric oxide synthase (iNOS) and NADPH oxidase (NOX) in RVSMCs. In addition, TA significantly decreased the production of tumor necrosis factor‑α (TNF‑α), interleukin 6 (IL‑6) and IL‑1β. Furthermore, TA pretreatment prevented nuclear factor‑κB (NF‑κB) subunit p65 phosphorylation and NF‑κB inhibitor α (IκBα) degradation induced by H2O2 in RVSMCs. TA is, therefore, suggested to inhibit H2O2-induced oxidative stress and inflammation in RVSMCs via inhibition of the NF‑κB signaling pathway. TA may have potential as a pharmacological agent in the prevention or treatment of atherosclerosis.
Sigma-Aldrich
Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-1 β ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Anti-NOX1, affinity isolated antibody