Merck
  • PKC-theta knockout mice are protected from fat-induced insulin resistance.
PKC-theta knockout mice are protected from fat-induced insulin resistance.
The Journal of clinical investigation (2004-09-17)
Jason K Kim, Jonathan J Fillmore, Mary Jean Sunshine, Bjoern Albrecht, Takamasa Higashimori, Dong-Wook Kim, Zhen-Xiang Liu, Timothy J Soos, Gary W Cline, William R O'Brien, Dan R Littman, Gerald I Shulman
ABSTRACT
Insulin resistance plays a primary role in the development of type 2 diabetes and may be related to alterations in fat metabolism. Recent studies have suggested that local accumulation of fat metabolites inside skeletal muscle may activate a serine kinase cascade involving protein kinase C-theta (PKC-theta), leading to defects in insulin signaling and glucose transport in skeletal muscle. To test this hypothesis, we examined whether mice with inactivation of PKC-theta are protected from fat-induced insulin resistance in skeletal muscle. Skeletal muscle and hepatic insulin action as assessed during hyperinsulinemic-euglycemic clamps did not differ between WT and PKC-theta KO mice following saline infusion. A 5-hour lipid infusion decreased insulin-stimulated skeletal muscle glucose uptake in the WT mice that was associated with 40-50% decreases in insulin-stimulated tyrosine phosphorylation of insulin receptor substrate-1 (IRS-1) and IRS-1-associated PI3K activity. In contrast, PKC-theta inactivation prevented fat-induced defects in insulin signaling and glucose transport in skeletal muscle. In conclusion, our findings demonstrate that PKC-theta is a crucial component mediating fat-induced insulin resistance in skeletal muscle and suggest that PKC-theta is a potential therapeutic target for the treatment of type 2 diabetes.
Sigma-Aldrich
D-Serine, ≥98% (TLC)
Sigma-Aldrich
PKCθ, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution