Merck
  • Quantification of 3-MCPD and its mercapturic metabolite in human urine: validation of an LC-MS-MS method and its application in the general population.
Quantification of 3-MCPD and its mercapturic metabolite in human urine: validation of an LC-MS-MS method and its application in the general population.
Analytical and bioanalytical chemistry (2015-04-12)
Roberta Andreoli, Martina Cirlini, Antonio Mutti
ABSTRACT
A new method for the simultaneous quantitative determination in human urine of 3-monochloropropane-1,2-diol (3-MCPD), a toxic food contaminant, and its metabolite, 2,3-dihydroxypropyl mercapturic acid (DHPMA), was developed and validated. After urine dilution, the analytes were separated on an Atlantis®dC18 column and quantified by liquid chromatography-tandem mass spectrometry using isotopically labelled internal standards. The limits of quantification (S/N = 10) were 1.90 and 2.21 μg/L for 3-MCPD and DHPMA, respectively. Intra- and inter-day precision were lower than 6 % for each compound. Matrix effects were evaluated. Due to the high sensitivity and good accuracy of the method, 3-MCPD and DHPMA were found in 67 and 100 % of urine samples of healthy subjects, respectively.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
(±)-3-Chloro-1,2-propanediol, 98%