Merck
  • R-Smad signaling-mediated VEGF expression coordinately regulates endothelial cell differentiation of rat mesenchymal stem cells.
R-Smad signaling-mediated VEGF expression coordinately regulates endothelial cell differentiation of rat mesenchymal stem cells.
Stem cells and development (2015-01-21)
Wen-Jia Ai, Jie Li, Shao-Mang Lin, Wen Li, Chi-Zhuai Liu, Wei-Ming Lv
ABSTRACT
A low-efficiency yield hinders the use of stem cells as a source of endothelial cells (ECs) for therapeutic vascularization, and the diversity of the transforming growth factor-β (TGF-β) superfamily has undermined understanding the effects of its potent vascularization-inducing. Herein, we studied the role of the TGF-β superfamily in EC differentiation of rat bone marrow mesenchymal stem cells (MSCs) induced by Smad2/3 and Smad1/5/8 signaling. MSCs that had been sorted by flow cytometry as CD31-negative were cultured for 14 days in medium supplemented with vascular endothelial growth factor (VEGF) and basic fibroblast growth factor (bFGF) as the control. The Smad2/3 pathway was activated by TGF-β1 and Smad1/5/8 by bone morphogenetic proteins (BMPs). In the early phase in the Smad2/3-activated group, there were 10% CD31-positive cells, which was significantly higher than in the control group. A low Smad1/5/8 phosphorylation level after BMP4 activation doubled the number of CD31-positive cells, while a higher phosphorylation level after BMP9 activation showed no effect. A Smad2/3 inhibitor initially blocked differentiation but later promoted it, while a Smad1/5/8 inhibitor reversed the induction observed with BMPs. Moreover, the positive effects of R-Smad on differentiation were weakened by the VEGF neutralizing antibody, and a Smad3 inhibitor decreased VEGF expression and blocked differentiation in both the early and late phases. In conclusion, differentiation of ECs from MSCs via Smad2/3 signaling is stage dependent. Activation, particularly by Smad3, significantly promotes differentiation at an early phase but later is suppressive. A low Smad1/5/8 phosphorylation level has a positive effect, and R-Smad effects are partly mediated by VEGF.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
GDF-2 human, recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Vascular Endothelial Growth Factor from mouse, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
SIS3, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining