Merck
  • Endothelial nitric oxide synthase gene intron 4 variable number tandem repeat polymorphism in β-thalassemia major: relation to cardiovascular complications.
Endothelial nitric oxide synthase gene intron 4 variable number tandem repeat polymorphism in β-thalassemia major: relation to cardiovascular complications.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis (2015-02-24)
Azza A G Tantawy, Amira A M Adly, Eman A Ismail, Shereen H Aly
ABSTRACT
Endothelial nitric oxide synthase (eNOS), an enzyme that generates nitric oxide, is a major determinant of endothelial function. Several eNOS gene polymorphisms have been reported as 'susceptibility genes' in various human diseases states, including cardiovascular, pulmonary and renal diseases. We studied the 27-base pair tandem repeat polymorphism in intron 4 of eNOS gene in 60 β-thalassemia major (β-TM) patients compared with 60 healthy controls and assessed its role in subclinical atherosclerosis and vascular complications. Patients were evaluated stressing on transfusion history, splenectomy, thrombotic events, echocardiography and carotid intima-media thickness (CIMT). Analysis of eNOS intron 4 gene polymorphism was performed by PCR. No significant difference was found between β-TM patients and controls with regard to the distribution of eNOS4 alleles or genotypes. The frequency of eNOS4a allele (aa and ab genotypes) was significantly higher in β-TM patients with pulmonary hypertension or cardiomyopathy. Logistic regression analysis revealed that eNOS4a allele was an independent risk factor for pulmonary hypertension in β-TM patients [odds ratio (OR) 2.2, 95% confidence interval (95% CI) 1.19-5.6; P < 0.001]. We suggest that eNOS intron 4 gene polymorphism is related to endothelial dysfunction and subclinical atherosclerosis and could be a possible genetic marker for prediction of increased susceptibility to cardiovascular complications.
Sigma-Aldrich
Brilliant Cresyl Blue ALD, Certified by the Biological Stain Commission
Sigma-Aldrich
Brilliant Cresyl Blue ALD, certified by the BSC
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder