Merck
  • Highly reproducible planar Sb₂S₃-sensitized solar cells based on atomic layer deposition.
Highly reproducible planar Sb₂S₃-sensitized solar cells based on atomic layer deposition.
Nanoscale (2014-10-29)
Dae-Hwan Kim, Sang-Ju Lee, Mi Sun Park, Jin-Kyu Kang, Jin Hyuck Heo, Sang Hyuk Im, Shi-Joon Sung
ABSTRACT
A high-quality Sb₂S₃ thin-absorber with controllable thickness was reproducibly formed by atomic layer deposition (ALD) technique. Compared with conventional chemical bath deposition (CBD), the Sb₂S₃ absorber deposited by ALD did not contain oxide or oxygen impurities and showed a very uniform thickness of Sb₂S₃ absorbers formed on a rough surface of dense blocking TiO₂/F-doped SnOv (bl-TiO₂/FTO) substrate. The planar ALD-Sb₂S₃ solar cells comprised of Au/Poly-3-hexylthiophene/ALD-Sb₂S₃/bl-TiO₂/FTO showed significantly improved power conversion efficiency of 5.77% at 1 sun condition and narrow efficiency deviation, whereas the planar CBD-Sb₂S₃ solar cells exhibited 2.17% power conversion efficiency. The high efficiency and good reproducibility of ALD-Sb₂S₃ solar cell devices is attributed to reduced backward recombination because of the inhibition of oxide defects within ALD-Sb₂S₃ absorber and the conformal deposition of very uniform Sb₂S₃ absorbers on the blocking TiO₂ surface by ALD process.
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sulfur-32S solution, IRMM®, certified reference material, Spike Isotopic Reference Material
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF