Merck
  • Effect of latent heat in boiling water on the synthesis of gold nanoparticles of different sizes by using the Turkevich method.
Effect of latent heat in boiling water on the synthesis of gold nanoparticles of different sizes by using the Turkevich method.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-11-14)
Wenchao Ding, Peina Zhang, Yijing Li, Haibing Xia, Dayang Wang, Xutang Tao
ABSTRACT
The Turkevich method, involving the reduction of HAuCl4 with citrate in boiling water, allows the facile production of monodisperse, quasispherical gold nanoparticles (AuNPs). Although, it is well-known that the size of the AuNPs obtained with the same recipe vary slightly (as little as approximately 4 nm), but noticeably, from one report to another, it has rarely been studied. The present work demonstrates that this size variation can be reconciled by the small, but noticeable, effect that the latent heat in boiling water has on the size of the AuNPs obtained by using the Turkevich method. The increase in latent heat during water boiling caused an approximately 3 nm reduction in the size of the as-prepared AuNPs; this reduction in size is mainly a result of accelerated nucleation driven by the extra heat. It was further demonstrated that, the heating temperature can be utilized as an additional measure to adjust the growth rate of AuNPs during the reduction of HAuCl4 with citrate in boiling water. Therefore, the latent heat of boiling solvents may provide one way to control nucleation and growth in the synthesis of monodisperse nanoparticles.
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)