Merck
  • Significance of osteopontin in the sensitivity of malignant pleural mesothelioma to pemetrexed.
Significance of osteopontin in the sensitivity of malignant pleural mesothelioma to pemetrexed.
International journal of oncology (2014-04-10)
Susumu Takeuchi, Masahiro Seike, Rintaro Noro, Chie Soeno, Teppei Sugano, Fenfei Zou, Haruka Uesaka, Nobuhiko Nishijima, Masaru Matsumoto, Yuji Minegishi, Kaoru Kubota, Akihiko Gemma
ABSTRACT
Pemetrexed (PEM) is currently recommended as one of the standard anticancer drugs for malignant pleural mesothelioma (MPM). However, the mechanism of the sensitivity of MPM to PEM remains unclear. We analyzed the antitumor effects of PEM in six MPM cell lines by MTS assay. To identify genes associated with drug sensitivity, we conducted gene expression profiling on the same set of cell lines using GeneChips and pathway analysis. Three cell lines were sensitive to PEM. A total fo 18 transcripts and 14 genes identified by GeneChips were significantly correlated with sensitivity to PEM. Pathway analysis revealed that osteopontin (SPP1/OPN) was an important target in PEM sensitivity. Overexpression of SPP1/OPN was observed in the sensitive cells by quantitative PCR and western blot analysis. Introduction of SPP1/OPN by lentiviral vector significantly enhanced the invasion activities of MPM cells. PEM treatment with SPP1/OPN knockdown inhibited the PEM-induced cell growth-inhibitory effect in PEM-sensitive cells. Expression of SPP1/OPN and AKT phosphorylation significantly decreased after PEM treatment of the PEM-sensitive cells. High immunohistochemical expression of SPP1/OPN was observed in two of three MPM patients who had a partial response to PEM-based chemotherapy. PEM has antitumor effects in MPM cells dependent on SPP1/OPN overexpression resulting in AKT activation. Our results suggest that SPP1 may be used as a single predictive biomarker of the effectiveness of PEM treatment in MPM.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sgpp1
Sigma-Aldrich
MISSION® esiRNA, targeting human SPP1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cxxc1
Sigma-Aldrich
MISSION® esiRNA, targeting human CXXC1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Spp1
Sigma-Aldrich
MISSION® esiRNA, targeting human SGPP1