Merck
  • Experimental evaluation of radioiodinated sennoside B as a necrosis-avid tracer agent.
Experimental evaluation of radioiodinated sennoside B as a necrosis-avid tracer agent.
Journal of drug targeting (2014-10-21)
Dongjian Zhang, Dejian Huang, Yun Ji, Cuihua Jiang, Yue Li, Meng Gao, Nan Yao, Xuejiao Liu, Haibo Shao, Su Jing, Yicheng Ni, Zhiqi Yin, Jian Zhang
ABSTRACT
Necrosis-avid agents are a class of compounds that selectively accumulate in the necrotic tissues after systemic administration, which can be used for in vivo necrosis imaging and targeted therapies. In order to search for a necrosis-avid tracer agent with improved drugability, we labelled iodine-131 on sennoside B (SB) as a naturally occurring median dianthrone compound. The necrosis targetability and clearance properties of (131)I-SB were evaluated in model rats with liver and muscle necrosis. On SPECT/CT images, a "hot spot" in the infarcted liver lobe and necrotic muscle was persistently observed at 24 h and 72 h post-injection (p.i.). Gamma counting of the tissues of interest revealed a radioactivity ratio of necrotic to viable liver at 4.6 and 3.4 and of necrotic to viable muscle at 7.0 and 8.8 at 24 h and 72 h p.i., respectively. The good match of autoradiographs and fluoromicroscopic images with corresponding histochemical staining suggested preferential uptake of (131)I-SB in necrotic tissue. Pharmacokinetic study revealed that (131)I-SB has an elimination half-life of 8.6 h. This study indicates that (131)I-SB shows not only prominent necrosis avidity but also favourable pharmacokinetics, which may serve as a potential necrosis-avid diagnostic agent for assessment of tissue viability.
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%