Merck
Hydrogen sulfide poisoning: an antidotal role for sodium nitrite?
Veterinary and human toxicology (1997-06-01)
A H Hall, B H Rumack
ABSTRACT
In 2 separate incidents, 6 patients were poisoned with hydrogen sulfide (H2S) in sewer gas. In the first incident, mixing acid- and sodium hydroxide-based drain cleaners in a confined space resulted in 4 poisonings and 2 deaths. Three would-be rescuers were seriously poisoned and 1 died. Two survivors had neurological sequelae. Sodium nitrite appeared to have some clinical efficacy in 1 case. The second incident involved 2 patients working on a pump in a sewage pond. A patient lying on a raft close to the pond surface was seriously poisoned; sodium nitrite was clinically efficacious and this patient survived without developing neurological sequelae. Sodium nitrite deserves further clinical study as a potential H2S antidote.
Product Description
Supelco
Sodium nitrite, analytical standard
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%