Merck
  • L-serine synthesis in the central nervous system: a review on serine deficiency disorders.
L-serine synthesis in the central nervous system: a review on serine deficiency disorders.
Molecular genetics and metabolism (2009-12-08)
L Tabatabaie, L W Klomp, R Berger, T J de Koning
ABSTRACT
The de novo synthesis of the amino acid L-serine plays an essential role in the development and functioning of the central nervous system (CNS). L-serine displays many metabolic functions during different developmental stages; among its functions providing precursors for amino acids, protein synthesis, nucleotide synthesis, neurotransmitter synthesis and L-serine derived lipids. Patients with congenital defects in the L-serine synthesizing enzymes present with severe neurological abnormalities and underscore the importance of this synthetic pathway. In this review, we will discuss the cellular functions of the L-serine pathway, structure and enzymatic properties of the enzymes involved and genetic defects associated with this pathway.
Product Description
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material