Merck
  • Neuropsychological speed tests and blood phenylalanine levels in patients with phenylketonuria: a meta-analysis.
Neuropsychological speed tests and blood phenylalanine levels in patients with phenylketonuria: a meta-analysis.
Neuroscience and biobehavioral reviews (2008-11-29)
Julia Albrecht, Sven F Garbade, Peter Burgard
ABSTRACT
Although pathogenesis of phenylketonuria is not completely understood, a low phenylalanine diet is effective to prevent severe neurological impairment, mental retardation and behavioural difficulties. Treatment recommendations heavily rely on neuropsychological research; however, single study results are ambiguous, what is reflected in substantial variation of US, British, German, Dutch and French recommendations for blood phenylalanine concentrations for adolescents and adults. We conducted a meta-analysis estimating the influence of age, phenylalanine level, and type of neuropsychological test on effect sizes (standardized differences between controls and patients) of computer-based speed measurements in phenylketonuric patients. The effect of blood phenylalanine level on effect size was more pronounced in children and adolescents than in adults, with choice reaction time being particularly sensitive for phenylalanine concentrations. Results corroborate all recommendations for children. With the exception of the US and Dutch recommendations, all recommendations for adolescents seem to be too liberal. The same effect size is predicted for adult phenylalanine concentrations between 750 and 1500mumol/L not suggesting a preference for any of the published treatment recommendations for adulthood.
Product Description
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Phenylalanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%