Merck
  • AlCl3-promoted formal [2 + 3]-cycloaddition of 1,1-cyclopropane diesters with N-benzylic sulfonamides to construct highly stereoselective indane derivatives.
AlCl3-promoted formal [2 + 3]-cycloaddition of 1,1-cyclopropane diesters with N-benzylic sulfonamides to construct highly stereoselective indane derivatives.
Organic letters (2014-03-19)
Mengyun Zhu, Jinqian Liu, Jianjun Yu, Liangshun Chen, Chunmei Zhang, Limin Wang
ABSTRACT
An unprecedented AlCl3-promoted formal [2 + 3]-cycloaddition of 1,1-cyclopropanes with readily available N-benzylic sulfonamides has been developed. Experimental evidence supports an unusual mechanism wherein the donor-acceptor cyclopropane serves as a source of 2-styrylmalonate rather than the "classical" 1,3-dipole. A broad range of 1,1-cyclopropanediesters undergo a carbocation-initiated cyclization reaction with N-benzylic sulfonamides to afford highly functionalized Indane derivatives in a fast and high-yielding procedure.
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminum chloride hydrate, 99.999% trace metals basis