Merck
  • Full evaporation dynamic headspace and gas chromatography-mass spectrometry for uniform enrichment of odor compounds in aqueous samples.
Full evaporation dynamic headspace and gas chromatography-mass spectrometry for uniform enrichment of odor compounds in aqueous samples.
Journal of chromatography. A (2012-05-01)
Nobuo Ochiai, Kikuo Sasamoto, Andreas Hoffmann, Kazunori Okanoya
ABSTRACT
A method for analysis of a wide range of odor compounds in aqueous samples at sub-ng mL⁻¹ to μg mL⁻¹ levels was developed by full evaporation dynamic headspace (FEDHS) and gas chromatography-mass spectrometry (GC-MS). Compared to conventional DHS and headspace solid phase microextraction (HS-SPME), FEDHS provides more uniform enrichment over the entire polarity range for odor compounds in aqueous samples. FEDHS at 80°C using 3 L of purge gas allows complete vaporization of 100 μL of an aqueous sample, and trapping and drying it in an adsorbent packed tube, while providing high recoveries (85-103%) of the 18 model odor compounds (water solubility at 25°C: log0.54-5.65 mg L⁻¹, vapor pressure at 25°C: 0.011-3.2 mm Hg) and leaving most of the low volatile matrix behind. The FEDHS-GC-MS method showed good linearity (r²>0.9909) and high sensitivity (limit of detection: 0.21-5.2 ng mL⁻¹) for the model compounds even with the scan mode in the conventional MS. The feasibility and benefit of the method was demonstrated with analyses of key odor compounds including hydrophilic and less volatile characteristics in beverages (whiskey and green tea). In a single malt whiskey sample, phenolic compounds including vanillin could be determined in the range of 0.92-5.1 μg mL⁻¹ (RSD<7.4%, n=6). For a Japanese green tea sample, 48 compounds including 19 potent odorants were positively identified from only 100 μL of sample. Heat-induced artifact formation for potent odorants was also examined and the proposed method does not affect the additional formation of thermally generated compounds. Eighteen compounds including 12 potent odorants (e.g. coumarin, furaneol, indole, maltol, and pyrazine congeners) were determined in the range of 0.21-110 ng mL⁻¹ (RSD<10%, n=6).
Product Description
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub