Merck
  • Rapid and sensitive liquid chromatography-tandem mass spectrometric method for the quantitation of metformin in human plasma.
Rapid and sensitive liquid chromatography-tandem mass spectrometric method for the quantitation of metformin in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-07-21)
Yingwu Wang, Yunbiao Tang, Jingkai Gu, J Paul Fawcett, Xu Bai
ABSTRACT
A rapid and sensitive liquid chromatography-tandem mass spectrometric (LC-MS-MS) method for the determination of metformin in human plasma using phenformin as internal standard has been developed and validated. Sample preparation of plasma involved acidification with acetic acid, deproteination with acetonitrile and washing with dichloromethane. Samples were then analyzed by HPLC on a short Nucleosil C18 column (5 microm, 50 mm x 4.6 mm i.d.) using a mobile phase consisting of acetonitrile:methanol:10mM ammonium acetate pH 7.0 (20:20:60, v/v/v) delivered at 0.65 ml/min. Detection was performed using an Applied Biosystems Sciex API 4000 mass spectrometer set at unit resolution in the multiple reaction monitoring (MRM) mode. Atmospheric pressure chemical ionization (APCI) was used for ion production. The assay was linear over the range 1-2000 ng/ml with intra- and inter-day precision of <8.6% and accuracy in the range 91-110%. The limit of detection was 250 pg/ml in plasma. The method was successfully applied to a clinical pharmacokinetic study of an extended-release tablet of metformin hydrochloride (500 mg) administered as a single oral dose.
Product Description
Supelco
Metformin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metformin hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Metformin hydrochloride, European Pharmacopoeia (EP) Reference Standard