Merck
  • Enantioselective oxidative damage of chiral pesticide dichlorprop to maize.
Enantioselective oxidative damage of chiral pesticide dichlorprop to maize.
Journal of agricultural and food chemistry (2011-03-16)
Tong Wu, Xiuying Li, Honglin Huang, Shuzhen Zhang
ABSTRACT
To investigate the enantioselective oxidative damage of the pesticide dichlorprop (DCPP) to maize, young seedlings were exposed to solutions of DCPP enantiomers and racemate at different concentrations. Early root development was more influenced by (R)-DCPP than racemic (rac)- and (S)-DCPP. Inhibition rates of seed germination, seedling biomass, and root and shoot elongation were all in the order of (R)-DCPP > (rac)-DCPP > (S)-DCPP treatments. The antioxidant enzyme activities of superoxide dismutase (SOD) and peroxidase (POD) were significantly upregulated by exposure to lower concentrations of (R)-DCPP than (rac)- and (S)-DCPP. Direct determination of the formation of hydroxyl radical (•OH) with electron paramagnetic resonance (EPR) spectroscopy indicated that the •OH level in maize roots followed the order of (R)-DCPP > (rac)-DCPP > (S)-DCPP treatments. All of these results provide solicited evidence of the significant enantioselective phytotoxicity of DCPP to maize with a higher toxicity of (R)-DCPP than (S)- and (rac)-DCPP.
Product Description
Supelco
Dichlorprop, PESTANAL®, analytical standard
Supelco
Dichloroprop-P, PESTANAL®, analytical standard