Merck
  • Inactivation and inhibition of African swine fever virus by monoolein, monolinolein, and gamma-linolenyl alcohol. Brief report.
Inactivation and inhibition of African swine fever virus by monoolein, monolinolein, and gamma-linolenyl alcohol. Brief report.
Archives of virology (1986-01-01)
A Sola, S Rodríguez, A García Gancedo, P Vilas, C Gil-Fernández
ABSTRACT
Monoolein, monolinolein and (most strongly) gamma-linolenyl alcohol inactivate ASF virus and inhibit its replication in Vero cells at 25 micrograms/ml while at 10 micrograms/ml no inactivation occurs but inhibition of replication in tissue culture is observed. This suggests two possibly different action mechanisms.
Sigma-Aldrich
1-Linoleoyl-rac-glycerol, ≥97%, liquid
Glycerol monolinoleate, European Pharmacopoeia (EP) Reference Standard