Merck
  • Photoluminescence studies of ZnO nanorods grown by plasma-assisted molecular beam epitaxy.
Photoluminescence studies of ZnO nanorods grown by plasma-assisted molecular beam epitaxy.
Journal of nanoscience and nanotechnology (2013-07-19)
Min Su Kim, Giwoong Nam, Jae-Young Leem
ABSTRACT
Metal catalyst-free ZnO nanorods were grown on PS with buffer layers grown at 450 degrees C by plasma-assisted molecular beam epitaxy. Room temperature and temperature-dependent photoluminescence were carried out to investigate the optical properties of the ZnO nanorods with the average diameter of 120 nm and length of 300 nm. Three emission peaks, free excition, neutral-donor exciton, and free electron-to-neutral acceptor, were observed at 10 K. Huang-Rhys factor S of the ZnO nanorods was 0.978, which is much higher than that of ZnO thin films. The values of Varshni's empirical equation fitting parameters were alpha = 4 x 10(-3) eV/K, beta = 4.1 x 10(4) K, and E9(0) = 3.388 eV and the activation energy was about 96 meV.
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Supelco
Zinc oxide, analytical standard