Merck
  • Optimization of growth conditions for the isolation of dextran producing Leuconostoc spp. from indigenous food sources.
Optimization of growth conditions for the isolation of dextran producing Leuconostoc spp. from indigenous food sources.
Pakistan journal of pharmaceutical sciences (2013-07-03)
Farwa Sarwat, Nuzhat Ahmed, Afsheen Aman, Shah Ali Ul Qader
ABSTRACT
Leuconostoc are known to produce dextran, which have great commercial importance in chemical, medical and food industry. The present study is an attempt to select the best medium for the isolation of indigenous dextran producing Leuconostoc, measuring their enzyme activities for dextransucrase, production of dextran and identification of dextran producing Leuconostoc CMG706, CMG707, CMG710 and CMG713. Since, dextran producing Leuconostoc produce slimy colonies, twenty-four slime producing bacterial strains were isolated from different food sources, fruits and vegetables. Three different isolation medium were evaluated for the isolation of Leuconostoc only and the best one was found to be one containing sucrose and sodium azide. Further, all slime producing bacterial strains were screened for enzyme activity of dextransucrase, which is responsible for dextran production. Four bacterial strains CMG706, CMG707, CMG710 and CMG713 giving high enzyme activities were selected for dextran production and identified.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 410,000
Supelco
Dextran, analytical standard, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 270,000
Supelco
Dextran, analytical standard, 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 5,000
Supelco
Dextran, analytical standard, 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 12,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 670,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran, analytical standard, 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 50,000
Supelco
Dextran, analytical standard, 50,000
Supelco
Dextran, analytical standard, 150,000