Merck
  • The PARACHUTE IV trial design and rationale: percutaneous ventricular restoration using the parachute device in patients with ischemic heart failure and dilated left ventricles.
The PARACHUTE IV trial design and rationale: percutaneous ventricular restoration using the parachute device in patients with ischemic heart failure and dilated left ventricles.
American heart journal (2013-03-30)
Marco A Costa, Michael Pencina, Serjan Nikolic, Thomas Engels, Barry Templin, William T Abraham
ABSTRACT
Left ventricle (LV) remodeling after anterior wall myocardial infarction leads to increased LV volumes, myocardial stress, and, ultimately, heart failure (HF). Patients have high morbidity and mortality risk, and treatment remains limited. Percutaneous ventricular restoration (PVR) therapy using the Parachute device, a fluoropolymer membrane stretched over a nitinol conical frame, is a novel approach to partition off the damaged myocardium. In the European and United States PARACHUTE feasibility trials, the observed rates of death or rehospitalization for HF were <17% at 12 months. These data compare favorably with historical data and support the need of a randomized trial to determine the clinical efficacy of PVR on outcomes for patients with ischemic HF. To determine the safety and efficacy of PVR utilizing a LV partitioning device, Parachute, in a randomized clinical trial compared with optimal medical therapy. This US pivotal trial is approved by the Food and Drug Administration (ClinicalTrials.gov Identifier: NCT01286116) and will randomly assign (1:1) 478 patients with New York Heart Association class III-IV ischemic HF, akinetic or dyskinetic LV wall abnormality, and ejection fraction between 15% and 35% to optimal medical therapy (control) versus Parachute device implantation in approximately 65 hospitals. The primary endpoint is death or rehospitalization for worsening HF. Sample size calculation assumes constant hazards and follow-up ≥12 months using an event-driven trial design. We reported the rational and design of the first multicenter randomized trial to test the efficacy of PVR using the Parachute device to treat patients with ischemic HF and dilated LV.
Sigma-Aldrich
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size