Merck
  • Blacks have a greater sensitivity to α1-adrenoceptor-mediated venoconstriction compared with whites.
Blacks have a greater sensitivity to α1-adrenoceptor-mediated venoconstriction compared with whites.
Hypertension (Dallas, Tex. : 1979) (2013-02-13)
Abiodun Adefurin, Laxmi V Ghimire, Utkarsh Kohli, Mordechai Muszkat, Gbenga G Sofowora, Sachin Y Paranjape, C Michael Stein, Daniel Kurnik
ABSTRACT
Blacks have increased hemodynamic responses to both physiological and pharmacological adrenergic stimulation compared with whites, and this may contribute to the greater prevalence of hypertension in this ethnic group. A small study suggested enhanced α1-adrenoreceptor-mediated arterial vasoconstriction in the forearm vasculature of blacks compared with whites, but it is unknown whether this reflects a generalized vascular phenomenon. The objective of this study was to examine the hypothesis that there are ethnic differences in venous α1-adrenoreceptor responsiveness. Using a linear variable differential transformer, we measured local dorsal hand vein responses to increasing doses of the selective α1-adrenoreceptor agonist, phenylephrine, in 106 subjects (64 whites and 42 blacks). There was wide interindividual variability in responses to phenylephrine. The dose that produced 50% of maximal constriction (ED50) ranged from 11 to 5442 ng/min, and maximal venoconstriction (Emax) ranged from 13.5% to 100%. Blacks (geometric mean ED50 =172 ng/min; 95% confidence interval, 115-256 ng/min) were more sensitive to phenylephrine than whites (310 ng/min; 95% confidence interval, 222-434 ng/min; unadjusted P=0.026; adjusted P=0.003). Median Emax was slightly higher in blacks (89%; interquartile range, 82% to 98%) compared with whites (85%; interquartile range, 75% to 95%; P=0.07). Taken together with previous findings in arterial vessels, our results suggest a generalized increased sensitivity to α1-adrenoreceptor-mediated vasoconstriction in blacks. Increased vascular α-adrenoreceptor sensitivity could predispose to hypertension, and future studies addressing the contribution of this mechanism to ethnic differences in the prevalence of hypertension will be of interest.
Product Description
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material