Merck
  • Supersaturatable formulations for the enhanced oral absorption of sirolimus.
Supersaturatable formulations for the enhanced oral absorption of sirolimus.
International journal of pharmaceutics (2013-02-12)
Min-Soo Kim, Jeong-Soo Kim, Wonkyung Cho, Kwang-Ho Cha, Hee-Jun Park, Junsung Park, Sung-Joo Hwang
ABSTRACT
The purpose of this study was to develop supersaturatable formulations for the enhanced solubility and oral absorption of sirolimus. Supersaturatable formulations of hydrophilic polymers and/or surfactants were screened by formulation screening, which is based on solvent casting. The solid dispersion particles in the optimized formulations were prepared by spray drying. The particles were characterized in vitro and in vivo. The most effective supersaturatable formulation found in the formulation screening process was hydroxypropylmethyl cellulose (HPMC)-D-α-tocopheryl polyethylene glycol 1000 succinate (TPGS), followed by HPMC-Sucroester. In addition, the supersaturated state generated from HPMC-TPGS and HPMC-Sucroester 15 particles prepared by spray drying significantly improved the oral absorption of sirolimus in rats. Based on the pharmacokinetic parameters and supporting in vitro supersaturated dissolution data, the enhanced supersaturation properties of sirolimus led to enhanced in vivo oral absorption. In addition, the experimental results from the formulation screening used in our study could be useful for enhancing the bioavailability of sirolimus in preformulation and formulation studies.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
D-α-Tocopherol polyethylene glycol 1000 succinate, BioXtra, water soluble vitamin E conjugate
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.