Merck
  • An allele of an ancestral transcription factor dependent on a horizontally acquired gene product.
An allele of an ancestral transcription factor dependent on a horizontally acquired gene product.
PLoS genetics (2013-01-10)
H Deborah Chen, Mollie W Jewett, Eduardo A Groisman
ABSTRACT
Changes in gene regulatory circuits often give rise to phenotypic differences among closely related organisms. In bacteria, these changes can result from alterations in the ancestral genome and/or be brought about by genes acquired by horizontal transfer. Here, we identify an allele of the ancestral transcription factor PmrA that requires the horizontally acquired pmrD gene product to promote gene expression. We determined that a single amino acid difference between the PmrA proteins from the human adapted Salmonella enterica serovar Paratyphi B and the broad host range S. enterica serovar Typhimurium rendered transcription of PmrA-activated genes dependent on the PmrD protein in the former but not the latter serovar. Bacteria harboring the serovar Typhimurium allele exhibited polymyxin B resistance under PmrA- or under PmrA- and PmrD-inducing conditions. By contrast, isogenic strains with the serovar Paratyphi B allele displayed PmrA-regulated polymyxin B resistance only when experiencing activating conditions for both PmrA and PmrD. We establish that the two PmrA orthologs display quantitative differences in several biochemical properties. Strains harboring the serovar Paratyphi B allele showed enhanced biofilm formation, a property that might promote serovar Paratyphi B's chronic infection of the gallbladder. Our findings illustrate how subtle differences in ancestral genes can impact the ability of horizontally acquired genes to confer new properties.
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications