Merck
  • Polymeric multilayers that localize the release of chlorhexidine from biologic wound dressings.
Polymeric multilayers that localize the release of chlorhexidine from biologic wound dressings.
Biomaterials (2012-07-13)
Ankit Agarwal, Tyler B Nelson, Patricia R Kierski, Michael J Schurr, Christopher J Murphy, Charles J Czuprynski, Jonathan F McAnulty, Nicholas L Abbott
ABSTRACT
Biologic wound dressings contain animal-derived components and are susceptible to high infection rates. To address this issue, we report an approach that permits incorporation of non-toxic levels of the small molecule antiseptic 'chlorhexidine' into biologic dressings. The approach relies on the fabrication of polyelectrolyte multilayer (PEMs) films containing poly(allylaminehydrochloride) (PAH), poly(acrylicacid) (PAA), and chlorhexidine acetate (CX) on elastomeric poly(dimethylsiloxane) (PDMS) sheets. The PEMs (20-100 nm thick) are subsequently stamped onto the wound-contact surface of a synthetic biologic dressing, Biobrane, which contains collagen peptides. Chlorhexidine loading in the PEMs was tailored by tuning the number of (CX/PAA) bilayers deposited, providing burst release of up to 0.98 ± 0.06 μg/cm(2) of CX over 24 h, followed by zero-order release of 0.35 ± 0.04 μg/cm(2)/day for another week. Although the CX concentrations released were below the reported in vitro cytotoxicity limit (5 μg/mL over 24 h) for human dermal fibroblasts, they killed 4 log(10) counts of pathogenic bacteria Staphylococcus aureus in solution. The CX/PEMs could be stamped onto Biobrane with high efficiency to provide CX release kinetics and in vitro antibacterial activity similar to that on PDMS stamps. In a full-thickness 'splinted' dermal wound-model in normal wild-type mice, the CX-functionalized Biobrane showed no decrease in either its adherence to the wound-bed or wound closure rate over 14 days. The murine wounds topically inoculated with ∼10(5) CFU/cm(2) of S. aureus and treated with CX-functionalized Biobrane demonstrated a 3 log(10) decrease in the wound's bacterial burden within 3 days, compared to persistent bacterial colonization found in wounds treated with unmodified Biobrane (n = 10 mice, p < 0.005). Overall, this study presents a promising approach to prevent bacterial colonization in wounds under biologic dressings.
Sigma-Aldrich
Poly(allylamine hydrochloride), average Mw 50,000
Sigma-Aldrich
Poly(allylamine) solution, average Mw ~65,000, 10 wt. % in H2O
Sigma-Aldrich
Poly(allylamine hydrochloride), average Mw ~17,500 (GPC vs. PEG std.)
Sigma-Aldrich
Poly(allylamine) solution, 20 wt. % in H2O