Merck
  • Characterization of diatomite and its application for the retention of radiocobalt: role of environmental parameters.
Characterization of diatomite and its application for the retention of radiocobalt: role of environmental parameters.
Journal of environmental radioactivity (2012-06-09)
Guodong Sheng, Huaping Dong, Yimin Li
ABSTRACT
Clay minerals have been extensively studied because of their strong sorption and complexation ability. In this work, diatomite was characterized by using acid-base titration. Retention of radionuclide (60)Co(II) from aqueous solution by sorption onto diatomite was investigated by using batch technique under various environmental conditions such as pH, ionic strength, humic acid (HA), fulvic acid (FA), and temperature. The results indicated that the sorption of Co(II) onto diatomite was strongly dependent on pH. At low pH value, the sorption of Co(II) was dominated by outer-sphere surface complexation and ion exchange with Na(+)/H(+) on diatomite surfaces, whereas inner-sphere surface complexation was the main sorption mechanism at high pH value. The D-R model fitted the sorption isotherms better than the Langmuir and Freundlich models. The thermodynamic parameters (ΔH(0), ΔS(0) and ΔG(0)) calculated from the temperature-dependent sorption isotherms suggested that the sorption of Co(II) was an endothermic and spontaneous process. In addition, diatomite showed higher sorption capacity than that of lots of the sorbents reported in the literatures we surveyed. From the results of Co(II) removal by diatomite, the optimum reaction conditions can be obtained for the maximum removal of Co(II) from water. It is clear that the best pH values of the system to remove Co(II) from solution by using diatomite are 7-8. Considering the low cost and effective disposal of Co(II)-contaminated wastewaters, the best condition for Co(II) removal is at room temperature and solid content of 0.5 g/L. The results might be important for assessing the potential of practical application of diatomite in Co(II) and related radionuclide pollution management.
Product Description
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined