Merck
  • Practical synthesis of N-Boc- and N-Cbz-α-amido stannanes from α-amido sulfones using TMSSnBu3 and CsF.
Practical synthesis of N-Boc- and N-Cbz-α-amido stannanes from α-amido sulfones using TMSSnBu3 and CsF.
Organic letters (2011-04-08)
Tsuyoshi Mita, Yuki Higuchi, Yoshihiro Sato
ABSTRACT
In the presence of TMSSnBu(3) and CsF, stannylation of N-Boc- and N-Cbz-α-amido sulfones proceeded very well to afford the corresponding α-amido stannanes in moderate-to-high yields. This reaction tolerated α-aryl-, alkenyl-, and alkyl-substituted α-amido sulfones as well as substrates containing either an ester or cyano moiety, which might be reactive with lithium or magnesium stannides employed in conventional stannylation.
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis