Merck
  • Peppermint oil decreases the production of virulence-associated exoproteins by Staphylococcus aureus.
Peppermint oil decreases the production of virulence-associated exoproteins by Staphylococcus aureus.
Molecules (Basel, Switzerland) (2011-02-18)
Jing Li, Jing Dong, Jia-Zhang Qiu, Jian-Feng Wang, Ming-Jing Luo, Hong-En Li, Bing-Feng Leng, Wen-Zhi Ren, Xu-Ming Deng
ABSTRACT
The present study aimed to evaluate the antimicrobial activity of peppermint oil against Staphylococcus aureus, and further investigate the influence of peppermint oil on S. aureus virulence-related exoprotein production. The data show that peppermint oil, which contained high contents of menthone, isomenthone, neomenthol, menthol, and menthyl acetate, was active against S. aureus with minimal inhibitory concentrations (MICs) ranging from 64-256 µg/mL, and the production of S. aureus exotoxins was decreased by subinhibitory concentrations of peppermint oil in a dose-dependent manner. The findings suggest that peppermint oil may potentially be used to aid in the treatment of S. aureus infections.
Sigma-Aldrich
Peppermint oil, natural, from Mentha piperita L.
Sigma-Aldrich
Peppermint oil, terpeneless, FCC
Sigma-Aldrich
Peppermint oil, purified by triple-distillation, FCC, FG
Sigma-Aldrich
Peppermint oil, natural, FCC, FG