Merck
Disulfide formation via sulfenamides.
Chemical communications (Cambridge, England) (2010-08-24)
Jia Pan, Ming Xian
ABSTRACT
A phosphine-mediated one-step disulfide formation from S-nitrosothiols has been developed. This reaction can convert unstable S-nitrosothiols to stable disulfides via sulfenamide intermediates under very mild conditions. It has the potential to be used for the detection of S-nitrosothiols.
Product Description
Supelco
Sulfamerazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%