Merck
  • Determination of non-steroidal anti-inflammatory drugs residues in animal muscles by liquid chromatography-tandem mass spectrometry.
Determination of non-steroidal anti-inflammatory drugs residues in animal muscles by liquid chromatography-tandem mass spectrometry.
Analytica chimica acta (2010-06-29)
Piotr Jedziniak, Teresa Szprengier-Juszkiewicz, Małgorzata Olejnik, Jan Zmudzki
ABSTRACT
A confirmatory method for the determination of residues of nine non-steroidal anti-inflammatory drugs and one metabolite in animal muscles has been developed. After enzymatic hydrolysis samples were extracted with acetonitrile and cleaned up using alumina and C(18) SPE cartridges. Liquid chromatography-tandem mass spectrometry was used for the separation and determination of analytes. The method was validated in bovine muscles, according to the Commission Decision 2002/657/EC criteria. Applicability of the method in the analysis of swine, horse and chicken muscles was checked by precision and recovery experiment. The influence of matrix effect on the quantification of non-steroidal anti-inflammatory drugs residues was investigated. The method was used for the confirmation of phenylbutazone and oxyphenbutazone in horse muscle sample.
Product Description
Supelco
Carprofen-d3, VETRANAL®, analytical standard
Sigma-Aldrich
Oxyphenbutazone, ≥98% (HPLC)