Merck
  • Determination of the leaching of polymeric ion-selective membrane components by stripping voltammetry.
Determination of the leaching of polymeric ion-selective membrane components by stripping voltammetry.
Talanta (2010-03-20)
Beata Paczosa-Bator, Robert Piech, Andrzej Lewenstam
ABSTRACT
This paper focuses on the quantitative determination of the loss of the components from plastic membranes of ion-selective electrodes (ISEs) during contact with aqueous bathing solutions. The leaching processes, which affect the ISE responses, are rarely characterized by independent methods. For this purpose, differential pulse cathodic stripping voltammetry (DP CSV) is used. This method, owing to its high sensitivity, acceptable recovery and accuracy, is a good tool to characterize the kinetics of leakage of the lipophilic salts. Sodium tetraphenylborate (NaTPB) leakage from the PVC-based sodium-selective membrane containing two different plasticizers, o-nitrophenyl octyl ether (o-NPOE) or di(2-ethylhexyl) sebacate (DOS) is presented. Correlation between the rate of leaching of the lipophilic salt and dielectric constants of the plasticizers is observed. The data obtained by DP CSV correlate well with potentiometric and electrochemical impedance responses. The observed outflow of TPB(-) is associated with decreasing potentiometric sensitivity to sodium and increasing bulk membrane resistance.
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)
Sigma-Aldrich
Ammonium tetraphenylborate, 99%
Sigma-Aldrich
Potassium tetraphenylborate, 97%