Merck
  • Reaction rate coefficients of OH radicals and Cl atoms with ethyl propanoate, n-propyl propanoate, methyl 2-methylpropanoate, and ethyl n-butanoate.
Reaction rate coefficients of OH radicals and Cl atoms with ethyl propanoate, n-propyl propanoate, methyl 2-methylpropanoate, and ethyl n-butanoate.
The journal of physical chemistry. A (2009-09-15)
Pablo M Cometto, Véronique Daële, Mahmoud Idir, Silvia I Lane, Abdelwahid Mellouki
ABSTRACT
Kinetics of the reactions of OH radicals and Cl atoms with four saturated esters have been investigated. Rate coefficients for the gas-phase reactions of OH radicals with ethyl propanoate (k(1)), n-propyl propanoate (k(2)), methyl 2-methylpropanoate (k(3)), and ethyl n-butanoate (k(4)) were measured using a conventional relative rate method and the pulsed laser photolysis-laser induced fluorescence technique. At (296 +/- 2) K, the rate coefficients obtained by the two methods were in good agreement. Significant curvatures in the Arrhenius plots have been observed in the temperature range 243-372 K for k(1), k(3), and k(4). The rate coefficients for the reactions of the four esters with Cl atoms were determined using the relative rate method at (296 +/- 2) K and atmospheric pressure. The values obtained are presented, compared with the literature values when they exist, and discussed. Reactivity trends and atmospheric implications for these esters are also presented.
Sigma-Aldrich
Ethyl butyrate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl butyrate, 99%
Sigma-Aldrich
Ethyl propionate, ≥97%, FCC, FG
Sigma-Aldrich
Ethyl propionate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Ethyl propionate, 99%
Supelco
Ethyl butyrate, analytical standard
Sigma-Aldrich
Ethyl butyrate, natural, ≥98%, FCC, FG