Merck
  • Evaluation of enteric matrix microspheres prepared by emulsion-solvent evaporation using scanning electron microscopy.
Evaluation of enteric matrix microspheres prepared by emulsion-solvent evaporation using scanning electron microscopy.
Journal of microencapsulation (2004-01-14)
W M Obeidat, J C Price
ABSTRACT
Theophylline microspheres were prepared by the emulsion-solvent evaporation method using cellulose acetate butyrate (CAB381-20) and mixtures of CAB381-20(R) and cellulose acetate phthalate. The physical state of the drug, polymers and microspheres surfaces were determined using scanning electron microscopy. For those microspheres prepared using mixtures of CAB381-20 and cellulose acetate phthalate, scanning electron micrographs were taken before dissolution and also at different stages of dissolution (in SGF, pH 1.2 and in simulated intestinal fluid, pH 7.5). Micrographs were taken of the outside surfaces of the microspheres and of the cleaved microspheres showing their interiors (core). Drug crystals were observed on or near the surface of microspheres prepared from the polymer mixtures, while no drug particles or crystals were seen on the surfaces of microspheres prepared solely from CAB381-20. An acid wash for less than 2 min was capable of extracting all drug on the surface of the microspheres prepared from a mixture of CAB381-20 and cellulose acetate phthalate. The absence of drug crystals on the surface of CAB381-20 microspheres is believed to prevent initial drug release and create a lag time in release profiles. Results suggest that in both microsphere formulations, a layer of drug-free polymer is formed outside the core matrix and is believed to be responsible for the near zero-order release profiles.
Sigma-Aldrich
Cellulose acetate butyrate
Sigma-Aldrich
Cellulose acetate butyrate, average Mn ~70,000
Sigma-Aldrich
Cellulose acetate butyrate, average Mn ~12,000
Sigma-Aldrich
Cellulose acetate butyrate, average Mn ~30,000
Sigma-Aldrich
Cellulose acetate butyrate, average Mn ~30,000