Merck
Photosensitive dermatitis due to sunscreen allergy in a child.
The Australasian journal of dermatology (2002-05-02)
Natasha Cook, Susanne Freeman
ABSTRACT
We report a case of photoallergic contact dermatitis to two sunscreen agents, methoxycinnamate and oxybenzone, occurring in a 6-year-old Asian boy.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%