Merck
  • Degradation of the coffee flavor compound furfuryl mercaptan in model Fenton-type reaction systems.
Degradation of the coffee flavor compound furfuryl mercaptan in model Fenton-type reaction systems.
Journal of agricultural and food chemistry (2002-04-04)
Imre Blank, Ederlinda C Pascual, Stéphanie Devaud, Laurent B Fay, Richard H Stadler, Chahan Yeretzian, Bernard A Goodman
ABSTRACT
The stability of the coffee flavor compound furfuryl mercaptan has been investigated in aqueous solutions under Fenton-type reaction conditions. The impact of hydrogen peroxide, iron, ascorbic acid, and ethylenediaminetetraacetic acid was studied in various combinations of reagents and temperature. Furfuryl mercaptan reacts readily under Fenton-type reaction conditions, leading to up to 90% degradation within 1 h at 37 degrees C. The losses were lower when one or more of the reagents was omitted or the temperature decreased to 22 degrees C. Volatile reaction products identified were mainly dimers of furfuryl mercaptan, difurfuryl disulfide being the major compound. In addition, a large number of nonvolatile compounds was observed with molecular masses in the range of 92-510 Da. The formation of hydroxyl and carbon-centered radicals was indicated by electron paramagnetic resonance spectra using alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone or 5-(diethoxyphosphoryl)-5-methyl-1-pyrroline-N-oxide as spin traps. Whereas *OH was generated by Fenton-type reactions, the C-centered radical is probably a secondary product of the reaction of *OH with various organic molecules, the reaction with furfuryl mercaptan appearing to be the most important. No evidence for S-centered radicals was seen in the spin-trapping experiments, but a sulfur-containing radical was detected when measurements were made at 77 K in the absence of spin traps.
Sigma-Aldrich
2-Furanmethanethiol, 98%
Sigma-Aldrich
Furfuryl mercaptan, natural, 98%, FG
Sigma-Aldrich
Furfuryl mercaptan, ≥97%, FG
Sigma-Aldrich
2,2′-(Dithiodimethylene)difuran, ≥95%, FG